Assay Details
| Gene Symbol: ARSL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097988.1 | NP_001091457.1 | 3 - 4 | 339 | 59 | |
| GenBank mRNA | BC140583.1 | - | 3 - 4 | 339 | 59 | |
| BT030685.1 | - | 3 - 4 | 445 | 59 | ||
More Information
Set Membership: |
|

ARSL
Genomic Map