Assay Details
| Gene Symbol: ARSL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097988.1 | NP_001091457.1 | 6 - 7 | 1013 | 84 | |
| GenBank mRNA | BC140583.1 | - | 6 - 7 | 1013 | 84 | |
| BT030685.1 | - | 6 - 7 | 1119 | 84 | ||
More Information
Set Membership: |
|

ARSL
Genomic Map