Assay Details
| Gene Symbol: TRMO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097994.2 | NP_001091463.1 | 1 - 2 | 125 | 83 | |
| GenBank mRNA | BC140694.1 | - | 1 - 2 | 127 | 83 | |
More Information
Set Membership: |
|

TRMO
Genomic Map