Assay Details
| Gene Symbol: GOLGA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098007.1 | NP_001091476.1 | 4 - 5 | 1132 | 68 | |
| GenBank mRNA | BC140485.1 | - | 4 - 5 | 1132 | 68 | |
More Information
Set Membership: |
|

GOLGA5
Genomic Map