Assay Details
| Gene Symbol: ATP6AP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098022.1 | NP_001091491.1 | 1 - 2 | 193 | 115 | |
| GenBank mRNA | BC140624.1 | - | 1 - 2 | 193 | 115 | |
More Information
Set Membership: |
|

ATP6AP2
Genomic Map