Assay Details
| Gene Symbol: AKAP10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098024.1 | NP_001091493.1 | 5 - 6 | 981 | 80 | |
| GenBank mRNA | BC140679.1 | - | 5 - 6 | 981 | 80 | |
| EU239246.1 | - | 5 - 6 | 970 | 80 | ||
More Information
Set Membership: |
|

AKAP10
Genomic Map