Assay Details
| Gene Symbol: PUM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098030.1 | NP_001091499.1 | 16 - 17 | 1771 | 105 | |
| GenBank mRNA | BC133332.1 | - | 16 - 17 | 1771 | 105 | |
| BT030563.1 | - | 16 - 17 | 1759 | 105 | ||
More Information
Set Membership: |
|

PUM3
Genomic Map