Assay Details
| Gene Symbol: PTGDR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098034.1 | NP_001091503.1 | 1 - 2 | 906 | 78 | |
| GenBank mRNA | BC140594.1 | - | 1 - 2 | 906 | 78 | |
More Information
Set Membership: |
|

PTGDR
Genomic Map