Assay Details
| Gene Symbol: CSRNP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098035.1 | NP_001091504.1 | 3 - 4 | 636 | 82 | |
| GenBank mRNA | BT030484.1 | - | 3 - 4 | 636 | 82 | |
More Information
Set Membership: |
|

CSRNP1
Genomic Map