Assay Details
| Gene Symbol: SH2D3C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098039.1 | NP_001091508.1 | 9 - 10 | 2183 | 57 | |
| GenBank mRNA | BC140599.1 | - | 9 - 10 | 2183 | 57 | |
More Information
Set Membership: |
|

SH2D3C
Genomic Map