Assay Details
| Gene Symbol: MAML2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098050.1 | NP_001091519.1 | 3 - 4 | 2477 | 82 | |
| GenBank mRNA | BC140537.1 | - | 3 - 4 | 2477 | 82 | |
More Information
Set Membership: |
|

MAML2
Genomic Map