Assay Details
Gene Symbol: TNFRSF11B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098056.2 | NP_001091525.1 | 3 - 4 | 793 | 61 | |
GenBank mRNA | BC140651.1 | - | 3 - 4 | 794 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
