Assay Details
| Gene Symbol: RPS7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098874.2 | NP_001092344.1 | 1 - 2 | 62 | 63 | |
| GenBank mRNA | BC146133.1 | - | 1 - 2 | 64 | 63 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC146133.1, NM_001098874.2 transcripts.
Set Membership: |
|

RPS7
Genomic Map