Assay Details
| Gene Symbol: RPS7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098874.2 | NP_001092344.1 | 5 - 6 | 422 | 107 | |
| GenBank mRNA | BC146133.1 | - | 5 - 6 | 424 | 107 | |
More Information
Set Membership: |
|

RPS7
Genomic Map