Assay Details
| Gene Symbol: BLOC1S5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098875.1 | NP_001092345.1 | 1 - 2 | 119 | 99 | |
| GenBank mRNA | BC146091.1 | - | 1 - 2 | 119 | 99 | |
More Information
Set Membership: |
|

BLOC1S5
Genomic Map