Assay Details
| Gene Symbol: DUSP19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098878.1 | NP_001092348.1 | 1 - 2 | 364 | 80 | |
| GenBank mRNA | BC146174.1 | - | 1 - 2 | 364 | 80 | |
More Information
Set Membership: |
|

DUSP19
Genomic Map