Assay Details
| Gene Symbol: HCCS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098886.1 | NP_001092356.1 | 689 | 60 | ||
| GenBank mRNA | BC142105.1 | - | 689 | 60 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HCCS| Gene Symbol: HCCS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098886.1 | NP_001092356.1 | 689 | 60 | ||
| GenBank mRNA | BC142105.1 | - | 689 | 60 | ||
Set Membership: |
|