Assay Details
| Gene Symbol: CILK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098887.1 | NP_001092357.1 | 7 - 8 | 1062 | 102 | |
| GenBank mRNA | BC142307.1 | - | 7 - 8 | 1062 | 102 | |
More Information
Set Membership: |
|

CILK1
Genomic Map