Assay Details
| Gene Symbol: COPS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098904.1 | NP_001092374.1 | 10 - 11 | 1196 | 58 | |
| GenBank mRNA | BC146183.1 | - | 10 - 11 | 1196 | 58 | |
More Information
Set Membership: |
|

COPS3
Genomic Map