Assay Details
| Gene Symbol: ZFAND2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098906.2 | NP_001092376.1 | 7 - 8 | 691 | 76 | |
| GenBank mRNA | BC146106.1 | - | 7 - 8 | 692 | 76 | |
More Information
Set Membership: |
|

ZFAND2B
Genomic Map