Assay Details
| Gene Symbol: TSR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098911.2 | NP_001092381.1 | 5 - 6 | 823 | 55 | |
| GenBank mRNA | BC142410.1 | - | 5 - 6 | 824 | 55 | |
More Information
Set Membership: |
|

TSR3
Genomic Map