Assay Details
| Gene Symbol: RPS19BP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098916.2 | NP_001092386.1 | 2 - 3 | 247 | 68 | |
| GenBank mRNA | BC146267.1 | - | 2 - 3 | 248 | 68 | |
More Information
Set Membership: |
|

RPS19BP1
Genomic Map