Assay Details
| Gene Symbol: USP10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098924.1 | NP_001092394.1 | 4 - 5 | 1273 | 70 | |
| GenBank mRNA | BC142223.1 | - | 4 - 5 | 1273 | 70 | |
More Information
Set Membership: |
|

USP10
Genomic Map