Assay Details
| Gene Symbol: SERPINB10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098925.1 | NP_001092395.1 | 7 - 8 | 841 | 75 | |
| GenBank mRNA | BC142142.1 | - | 7 - 8 | 841 | 75 | |
More Information
Set Membership: |
|

SERPINB10
Genomic Map