Assay Details
| Gene Symbol: RNF135 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098936.1 | NP_001092406.1 | 2 - 3 | 511 | 90 | |
| GenBank mRNA | BC146080.1 | - | 2 - 3 | 511 | 90 | |
More Information
Set Membership: |
|

RNF135
Genomic Map