Assay Details
| Gene Symbol: UGT2A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098944.1 | NP_001092414.1 | 5 - 6 | 1321 | 99 | |
| GenBank mRNA | BC146247.1 | - | 5 - 6 | 1321 | 99 | |
More Information
Set Membership: |
|

UGT2A1
Genomic Map