Assay Details
| Gene Symbol: SEPTIN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098947.2 | NP_001092417.1 | 2 - 3 | 140 | 89 | |
| GenBank mRNA | BC142247.1 | - | 2 - 3 | 141 | 89 | |
More Information
Set Membership: |
|

SEPTIN1
Genomic Map