Assay Details
| Gene Symbol: NEK6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098988.2 | NP_001092458.1 | 6 - 7 | 671 | 57 | |
| GenBank mRNA | BC146269.1 | - | 6 - 7 | 673 | 57 | |
More Information
Set Membership: |
|

NEK6
Genomic Map