Assay Details
| Gene Symbol: SERPINA10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098997.2 | NP_001092467.1 | 1 - 2 | 45 | 84 | |
| GenBank mRNA | BC141987.1 | - | 1 - 2 | 46 | 84 | |
| BC149426.1 | - | 1 - 2 | 60 | 84 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC141987.1, NM_001098997.2 transcripts.
Set Membership: |
|

SERPINA10
Genomic Map