Assay Details
| Gene Symbol: SERPINA10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098997.2 | NP_001092467.1 | 2 - 3 | 827 | 114 | |
| GenBank mRNA | BC141987.1 | - | 2 - 3 | 828 | 114 | |
More Information
Set Membership: |
|

SERPINA10
Genomic Map