Assay Details
| Gene Symbol: NFS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099001.1 | NP_001092471.1 | 3 - 4 | 374 | 58 | |
| GenBank mRNA | BC142478.1 | - | 3 - 4 | 374 | 58 | |
More Information
Set Membership: |
|

NFS1
Genomic Map