Assay Details
| Gene Symbol: ORAI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099002.1 | NP_001092472.1 | 1 - 2 | 548 | 76 | |
| GenBank mRNA | BC142419.1 | - | 1 - 2 | 548 | 76 | |
More Information
Set Membership: |
|

ORAI1
Genomic Map