Assay Details
| Gene Symbol: BRAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099008.1 | NP_001092478.1 | 1272 | 69 | ||
| GenBank mRNA | BC146079.1 | - | 1272 | 69 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

BRAP| Gene Symbol: BRAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099008.1 | NP_001092478.1 | 1272 | 69 | ||
| GenBank mRNA | BC146079.1 | - | 1272 | 69 | ||
Set Membership: |
|