Assay Details
| Gene Symbol: MTREX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099012.1 | NP_001092482.1 | 5 - 6 | 540 | 64 | |
| GenBank mRNA | BC142299.1 | - | 5 - 6 | 540 | 64 | |
| BC146077.1 | - | 5 - 6 | 691 | 64 | ||
More Information
Set Membership: |
|

MTREX
Genomic Map