Assay Details
| Gene Symbol: PCDHGA8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099705.1 | NP_001093175.1 | 1 - 2 | 2583 | 116 | |
| GenBank mRNA | BC142514.1 | - | 1 - 2 | 2583 | 116 | |
More Information
Set Membership: |
|

PCDHGA8
Genomic Map