Assay Details
| Gene Symbol: CLIC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100301.2 | NP_001093771.1 | 1 - 2 | 58 | 100 | |
| GenBank mRNA | BC147880.1 | - | 1 - 2 | 59 | 100 | |
More Information
Set Membership: |
|

CLIC3
Genomic Map