Assay Details
| Gene Symbol: FCER1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100310.2 | NP_001093780.1 | 3 - 4 | 386 | 86 | |
| GenBank mRNA | BC149946.1 | - | 3 - 4 | 387 | 86 | |
More Information
Set Membership: |
|

FCER1A
Genomic Map