Assay Details
| Gene Symbol: PIGL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100311.2 | NP_001093781.1 | 6 - 7 | 674 | 84 | |
| GenBank mRNA | BC149604.1 | - | 6 - 7 | 676 | 84 | |
More Information
Set Membership: |
|

PIGL
Genomic Map