Assay Details
| Gene Symbol: EID3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100312.1 | NP_001093782.1 | 1 | 1146 | 96 | |
| GenBank mRNA | BC149257.1 | - | 1 | 1146 | 96 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

EID3| Gene Symbol: EID3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100312.1 | NP_001093782.1 | 1 | 1146 | 96 | |
| GenBank mRNA | BC149257.1 | - | 1 | 1146 | 96 | |
Set Membership: |
|