Assay Details
| Gene Symbol: DHRS12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100313.1 | NP_001093783.1 | 7 - 8 | 651 | 57 | |
| GenBank mRNA | BC149086.1 | - | 7 - 8 | 651 | 57 | |
More Information
Set Membership: |
|

DHRS12
Genomic Map