Assay Details
| Gene Symbol: TMEM189 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100315.1 | NP_001093785.1 | 1 - 2 | 152 | 48 | |
| GenBank mRNA | BC148012.1 | - | 1 - 2 | 152 | 48 | |
| BC149405.1 | - | 1 - 2 | 191 | 48 | ||
More Information
Set Membership: |
|

TMEM189
Genomic Map