Assay Details
Gene Symbol: C22H3orf62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100318.1 | NP_001093788.1 | 1 - 2 | 649 | 78 | |
GenBank mRNA | BC149382.1 | - | 1 - 2 | 649 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
