Assay Details
| Gene Symbol: GALNT12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100325.1 | NP_001093795.1 | 9 - 10 | 1614 | 99 | |
| GenBank mRNA | BC148121.1 | - | 9 - 10 | 1614 | 99 | |
More Information
Set Membership: |
|

GALNT12
Genomic Map