Assay Details
| Gene Symbol: RIPK4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100327.1 | NP_001093797.1 | 2 - 3 | 472 | 51 | |
| GenBank mRNA | BC148094.1 | - | 2 - 3 | 472 | 51 | |
More Information
Set Membership: |
|

RIPK4
Genomic Map