Assay Details
| Gene Symbol: TOMM22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100335.1 | NP_001093805.1 | 2 - 3 | 236 | 80 | |
| GenBank mRNA | BC149339.1 | - | 2 - 3 | 236 | 80 | |
| HM347496.1 | - | 2 - 3 | 227 | 80 | ||
More Information
Set Membership: |
|

TOMM22
Genomic Map