Assay Details
| Gene Symbol: PRADC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100338.2 | NP_001093808.1 | 1 - 2 | 151 | 96 | |
| GenBank mRNA | BC150018.1 | - | 1 - 2 | 152 | 96 | |
More Information
Set Membership: |
|

PRADC1
Genomic Map