Assay Details
| Gene Symbol: SRP14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100350.1 | NP_001093820.1 | 3 - 4 | 275 | 134 | |
| GenBank mRNA | BC149893.1 | - | 3 - 4 | 275 | 134 | |
More Information
Set Membership: |
|

SRP14
Genomic Map