Assay Details
| Gene Symbol: ZNF391 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100352.1 | NP_001093822.1 | 2 - 3 | 373 | 118 | |
| GenBank mRNA | BC149512.1 | - | 2 - 3 | 373 | 118 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC149512.1, NM_001100352.1 transcripts.
Set Membership: |
|

ZNF391
Genomic Map