Assay Details
| Gene Symbol: SYNGR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100358.1 | NP_001093828.1 | 3 - 4 | 487 | 85 | |
| GenBank mRNA | BT030736.1 | - | 3 - 4 | 487 | 85 | |
More Information
Set Membership: |
|

SYNGR2
Genomic Map