Assay Details
| Gene Symbol: GPD1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100363.2 | NP_001093833.1 | 1 - 2 | 137 | 99 | |
| GenBank mRNA | BC149966.1 | - | 1 - 2 | 138 | 99 | |
More Information
Set Membership: |
|

GPD1L
Genomic Map